CAS RN 169213-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol, TRC
CAS: 169213-73-4 Molecular Formula: C5H5N2OF3 Molecular Weight (g/mol): 166.1 Synonym: 5-(Trifluoromethyl)-1H-pyrazole-3-methanol,(5-Trifluoromethyl-2H-pyrazol-3-yl)-methanol,[5-(Trifluoromethyl)-1H-pyrazol-3-yl]methanol IUPAC Name: [3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol SMILES: OCc1cc(n[nH]1)C(F)(F)F