missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrid-3-ylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10452913
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10452913 250 mg 250mg
10507103 1 g 1g
10429094 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10452913 Supplier Thermo Scientific Maybridge Supplier No. CC39801CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4385-77-7
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03426512
InChI Key PXASTGBSGPFLFJ-UHFFFAOYSA-N
Synonym 3-pyridin-3-yl-benzoic acid, 3-pyridin-3-yl benzoic acid, 3-3'-pyridyl benzoic acid, 3-3-pyridinyl benzoic acid, 3-pyrid-3-ylbenzoic acid, 3-3-pyridyl benzoic acid, benzoic acid, 3-3-pyridinyl, acmc-1bmxh, 3-3'-pyridyl benzoicacid, 3-3-carboxyphenyl pyridine
PubChem CID 2795575
IUPAC Name 3-pyridin-3-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CN=CC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 3-Pyrid-3-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.