CAS RN 4385-77-7
IUPAC Name:
3-(pyridin-3-yl)benzoate
Synonyms:
3-(pyridin-3-yl)benzoate
Molecular Weight (g/mol):
198.2
Molecular Formula:
C12H8NO2
InChi Key:
PXASTGBSGPFLFJ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=CC(=C1)C1=CC=CN=C1
1
–
1
of
1
results
1
3-Pyrid-3-ylbenzoic acid, 95%, Thermo Scientific™
CAS: 4385-77-7 Molecular Formula: C12H9NO2 Molecular Weight (g/mol): 199.209 MDL Number: MFCD03426512 InChI Key: PXASTGBSGPFLFJ-UHFFFAOYSA-N Synonym: 3-pyridin-3-yl-benzoic acid,3-pyridin-3-yl benzoic acid,3-3'-pyridyl benzoic acid,3-3-pyridinyl benzoic acid,3-pyrid-3-ylbenzoic acid,3-3-pyridyl benzoic acid,benzoic acid, 3-3-pyridinyl,acmc-1bmxh,3-3'-pyridyl benzoicacid,3-3-carboxyphenyl pyridine PubChem CID: 2795575 IUPAC Name: 3-pyridin-3-ylbenzoic acid SMILES: C1=CC(=CC(=C1)C(=O)O)C2=CN=CC=C2