missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 3-Prong Retort Clamp
Bochem™ 3-Prong Retort Clamp with Stainless Steel construction and silicone coated fingers.
Brand: Bochem™ 5420
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Stainless Steel
- 3-prong
- Silicone coated fingers
Specifications
Stainless Steel (18/10) | |
190 mm | |
3-Prong |
12 mm | |
0 to 120 mm |