missing translation for 'onlineSavingsMsg'
Learn More

3-Octanone, 98%

Product Code. 11480043
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480043 500 g 500g
11460043 25 g 25g
11470043 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480043 Supplier Thermo Scientific Alfa Aesar Supplier No. A12113.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 106-68-3 | C8H16O | 128.215 g/mol

3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-68-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009515
InChI Key RHLVCLIPMVJYKS-UHFFFAOYSA-N
Synonym 3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural
PubChem CID 246728
ChEBI CHEBI:80946
IUPAC Name octan-3-one
SMILES CCCCCC(=O)CC

Specifications

Melting Point -23°C
Density 0.822
Boiling Point 167°C to 168°C
Flash Point 51°C (123°F)
Refractive Index 1.415
Quantity 500 g
UN Number UN1224
Beilstein 1700021
Merck Index 14,6084
Solubility Information Insoluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 3-Octanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.