Learn More
Eppendorf™ Research 3 Neo Piston Mount
Product Code. 30689645 Shop All Eppendorf Products
Click to view available options
Type:
Piston Mount
Unit Size:
Pack of 5
Product Code. 30689645
Brand: Eppendorf™ 3102631000
The piston mount secures the piston, ensuring smooth and accurate movement
  • Replacing a worn or damaged mount restores the pipette's performance
  • Ensuring continued precision in laboratory work
  • The piston mount secures the piston
  • Ensuring smooth and accurate movement, which is crucial for precise volume control
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Plastic
Type Piston Mount
For Use With (Equipment) 3 Neo, 2.5 to 1000 μL
Quantity 5 Pk.