missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research 3 Neo Pipette Carousel and Holder

Product Code. p-200135750 Shop All Eppendorf Products
Click to view available options
Product Type:
Pipette Carousel
Pipette Holder
Positions:
1
6
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30689724

Brand: Eppendorf™ 3116000236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments

  • The Eppendorf Pipette Holder System stores all current mechanical and electronic Eppendorf pipettes and most of their predecessors
  • Pipette Carousel 2, white Rotatable holder with 6 holders for Eppendorf Research™ 3 neo pipettes
  • Carousels and Stands with interchangeable pipette holders and charger shells, no tools required
  • Large rubber feet protect carousels and stands from liquids spilled on bench top\n
  • Pictograms on all charger shells and pipette holders for clear assignment in the lab
  • Save valuable bench space with the Eppendorf Pipette Holder System
  • With space for up to six single- or multi-channel pipettes, you can easily store all of your Eppendorf pipettes and multi-dispensers within easy reach
  • Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors
TRUSTED_SUSTAINABILITY

Tekniske data

Product Type Pipette Carousel
Autoclavable Not Autoclavable
Color White
Holds Manual Pipettes (6)
Material Polycarbonate
Positions 6
Shape Circular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) 3 Neo
Quantity 1 Ea.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado