missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research 3 Neo Pipette Carousel and Holder

Product Code. 30689724 Shop All Eppendorf Products
Change view
Click to view available options
Product Type:
Pipette Carousel
Pipette Holder
Positions:
1
6
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Positions Unit Size
30689724 Pipette Carousel 6 Each
30689699 Pipette Holder 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30689724 Supplier Eppendorf™ Supplier No. 3116000236
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments

  • The Eppendorf Pipette Holder System stores all current mechanical and electronic Eppendorf pipettes and most of their predecessors
  • Pipette Carousel 2, white Rotatable holder with 6 holders for Eppendorf Research™ 3 neo pipettes
  • Carousels and Stands with interchangeable pipette holders and charger shells, no tools required
  • Large rubber feet protect carousels and stands from liquids spilled on bench top\n
  • Pictograms on all charger shells and pipette holders for clear assignment in the lab
  • Save valuable bench space with the Eppendorf Pipette Holder System
  • With space for up to six single- or multi-channel pipettes, you can easily store all of your Eppendorf pipettes and multi-dispensers within easy reach
  • Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color White
Holds Manual Pipettes (6)
Material Polycarbonate
Positions 6
Shape Circular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 3 Neo
Quantity 1 Ea.
Product Type Pipette Carousel
Shelves None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.