missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propyl mercaptoacetate, 98%, Thermo Scientific™

Product Code. 15486097
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 15486097

Brand: Thermo Scientific Alfa Aesar H33285.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 852997-30-9
Molecular Formula C6H12O2S2
Molecular Weight (g/mol) 180.28
MDL Number MFCD19440668
InChI Key CNFYMFGPHSETCO-UHFFFAOYSA-N
Synonym 3-methylthio propyl mercaptoacetate, unii-3ri9z9ir02, mercaptoacetic acid 3-methylthio propyl ester, fema no. 4561, acetic acid, mercapto-, 3-methylthio propyl ester, acetic acid, 2-mercapto-, 3-methylthio propyl ester, 3-methylsulfanyl propyl sulfanylacetate acetic acid, mercapto-,3-methylthio propyl ester, 3-methylsulfanyl propyl 2-sulfanylacetate
PubChem CID 46779104
IUPAC Name 3-methylsulfanylpropyl 2-sulfanylacetate
SMILES CSCCCOC(=O)CS

Specifications

Density 1.152
Boiling Point 112°C to 114°C (0.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula HSCH2CO2(CH2)3SCH3
Refractive Index 1.514
Quantity 50 g
Formula Weight 180.29
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propyl mercaptoacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.