CAS RN 852997-30-9
CAS RN 852997-30-9
IUPAC Name:
3-(methylsulfanyl)propyl 2-sulfanylacetate
Synonyms:
3-(methylsulfanyl)propyl 2-sulfanylacetate
Molecular Weight (g/mol):
180.28
Molecular Formula:
C6H12O2S2
InChi Key:
CNFYMFGPHSETCO-UHFFFAOYSA-N
SMILES:
CSCCCOC(=O)CS
1
–
1
of
1
results
1
3-(Methylthio)propyl mercaptoacetate, 98%, Thermo Scientific™
CAS: 852997-30-9 Molecular Formula: C6H12O2S2 Molecular Weight (g/mol): 180.28 MDL Number: MFCD19440668 InChI Key: CNFYMFGPHSETCO-UHFFFAOYSA-N Synonym: 3-methylthio propyl mercaptoacetate,unii-3ri9z9ir02,mercaptoacetic acid 3-methylthio propyl ester,fema no. 4561,acetic acid, mercapto-, 3-methylthio propyl ester,acetic acid, 2-mercapto-, 3-methylthio propyl ester,3-methylsulfanyl propyl sulfanylacetate acetic acid, mercapto-,3-methylthio propyl ester,3-methylsulfanyl propyl 2-sulfanylacetate PubChem CID: 46779104 IUPAC Name: 3-methylsulfanylpropyl 2-sulfanylacetate SMILES: CSCCCOC(=O)CS