missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhexanoic acid, 97%, Thermo Scientific™

Product Code. 15462172 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15462172

Brand: Thermo Scientific Acros 456600010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3780-58-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
InChI Key NZQMQVJXSRMTCJ-UHFFFAOYSA-N
Synonym 3-methylcaproic acid, 3-methyl-hexanoic acid, hexanoic acid, 3-methyl, 3-methylhexylic acid, 3-methyl hexanoic acid, hexanoicacid,3-methyl, 3-methyl-2-hexanoic acid, 2-02-00-00298 beilstein handbook reference
PubChem CID 95315
IUPAC Name 3-methylhexanoic acid
SMILES CCCC(C)CC(=O)O

Specifications

Boiling Point 215.0°C to 216.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Acid)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 3-Methylhexanoic acid
compliance-icons
Product Identifier
  • 3-Methylhexanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.