accessibility menu, dialog, popup

UserName

CAS RN 3780-58-3

H3CCH3OHO

CAS RN 3780-58-3

IUPAC Name: 3-methylhexanoic acid
Synonyms: 3-methylhexanoic acid
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: NZQMQVJXSRMTCJ-UHFFFAOYNA-N
SMILES: CCCC(C)CC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results