missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcinnamic acid, 98%, Thermo Scientific™

Product Code. 10451681 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 10451681

Brand: Thermo Scientific Acros 296940100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3029-79-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 116°C to 119°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H10O2
Linear Formula CH3C6H4CH=CHCO2H
MDL Number MFCD00016844
Quantity 10 g
Synonym 3-methylcinnamic acid, m-methylcinnamic acid, e-3-m-tolyl acrylic acid, 3-m-tolyl acrylic acid, 2e-3-3-methylphenyl acrylic acid, 2e-3-3-methylphenyl prop-2-enoic acid, cinnamic acid, m-methyl, 3-methylcinnamic acid, predominantly trans, 2e-3-3-methylphenyl-2-propenoic acid, 2-propenoic acid, 3-3-methylphenyl-, 2e
InChI Key JZINNAKNHHQBOS-AATRIKPKSA-N
SMILES CC1=CC=CC(=C1)C=CC(=O)O
IUPAC Name (E)-3-(3-methylphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 162.19
PubChem CID 2063448
ChEBI CHEBI:61118
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylcinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.