missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethyl)benzonitrile, 97%

Product Code. 11310136
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310136 10 g 10g
11320136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310136 Supplier Thermo Scientific Alfa Aesar Supplier No. H32271.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 261952-04-9 | C9H6F3N | 185.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261952-04-9
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD01631595
InChI Key CSJHJNBBWUGJFY-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethyl benzonitrile, benzonitrile,3-methyl-5-trifluoromethyl, 3-cyano-5-methylbenzotrifluoride, 3-methyl-5-trifluoromethyl-benzonitrile, 3-methyl-5-trifluoromethyl benzenecarbonitrile, 5-methyl-3-trifluoromethyl benzenecarbonitrile
PubChem CID 2775595
IUPAC Name 3-methyl-5-(trifluoromethyl)benzonitrile
SMILES CC1=CC(=CC(=C1)C#N)C(F)(F)F

Specifications

Refractive Index 1.463
Quantity 10 g
UN Number UN3276
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.