accessibility menu, dialog, popup

UserName

CAS RN 261952-04-9

IUPAC Name: 3-methyl-5-(trifluoromethyl)benzonitrile
Synonyms: 3-methyl-5-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: CSJHJNBBWUGJFY-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results