missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15455157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455157 1 g 1g
15465157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455157 Supplier Thermo Scientific Alfa Aesar Supplier No. H32444.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-55-8
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD08741403
InChI Key WRJCQRHFOGJLLM-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-methyl-5-trifluoromethoxy benzene, 3-bromomethyl-5-trifluoromethoxy toluene
PubChem CID 26985614
IUPAC Name 1-(bromomethyl)-3-methyl-5-(trifluoromethoxy)benzene
SMILES CC1=CC(=CC(=C1)OC(F)(F)F)CBr

Specifications

Refractive Index 1.482
Quantity 1 g
UN Number UN3265
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethoxy)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.