accessibility menu, dialog, popup

UserName

CAS RN 916420-55-8

CH3OFFFBr

CAS RN 916420-55-8

IUPAC Name: 1-(bromomethyl)-3-methyl-5-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-3-methyl-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: WRJCQRHFOGJLLM-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results