missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™

Product Code. 10182994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182994 800 mL AcroSeal™ Glass Bottle 800mL
10172994 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182994 Supplier Thermo Scientific Acros Supplier No. 426758000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36282-40-3 | C7H7BrMgO | 211.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36282-40-3 , 109-99-9 , 108-88-3
Molecular Formula C7H7BrMgO
Molecular Weight (g/mol) 211.34
MDL Number MFCD00672002
InChI Key AJKNYEHIJAMGHX-UHFFFAOYSA-M
Synonym 3-methoxyphenylmagnesium bromide, 3-methoxyphenyl magnesium bromide, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromide, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, 3-methoxyphenyl-magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide, grignard reagent
PubChem CID 4605969
SMILES COC1=CC=CC([Mg]Br)=C1

Specifications

CAS Min % 19.02, 76.75
CAS Max % 23.25, 80.98
Color Yellow to Brown
Density 1.0000g/mL
Flash Point −21°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3OC6H4MgBr
Quantity 800 mL
UN Number 2924
Specific Gravity 1
IUPAC Name magnesium(2+) anisole bromide
Formula Weight 211.34
Physical Form Solution
Chemical Name or Material 3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.