missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycinnamic acid, predominantly trans, 98+%

Product Code. 11479582
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479582 25 g 25g
11469582 5 g 5g
11489582 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479582 Supplier Thermo Scientific Alfa Aesar Supplier No. A10723.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1533748 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1533748
MDL Number MFCD00004386

Specifications

Melting Point 117°C to 121°C
Molecular Formula C10H10O3
Quantity 25 g
Synonym 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid
InChI Key LZPNXAULYJPXEH-AATRIKPKSA-N
SMILES COC1=CC=CC(=C1)C=CC(=O)O
IUPAC Name (E)-3-(3-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 178.19
PubChem CID 637668
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Methoxycinnamic acid, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.