missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-N-methylbenzylamine, 97%

Product Code. 15451098
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451098 1 g 1g
15461098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451098 Supplier Thermo Scientific Alfa Aesar Supplier No. H54970.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41789-95-1 | C9H14NO | 152.22 g/mol

3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41789-95-1
Molecular Formula C9H14NO
Molecular Weight (g/mol) 152.22
MDL Number MFCD04618244
InChI Key FIFKRPFWLHBMHL-UHFFFAOYSA-O
Synonym 3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine
PubChem CID 5152225
IUPAC Name 1-(3-methoxyphenyl)-N-methylmethanamine
SMILES C[NH2+]CC1=CC=CC(OC)=C1

Specifications

Density 1.014
Boiling Point 130°C to 133°C
Flash Point 100°C (212°F)
Refractive Index 1.529
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethyl acetate and methanol.
Formula Weight 151.21
Percent Purity 97%
Chemical Name or Material 3-Methoxy-N-methylbenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.