accessibility menu, dialog, popup

UserName

CH3NH2OCH3

CAS RN 41789-95-1

IUPAC Name: [(3-methoxyphenyl)methyl](methyl)azanium
Synonyms: [(3-methoxyphenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 152.22
Molecular Formula: C9H14NO
InChi Key: FIFKRPFWLHBMHL-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=CC(OC)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results