Learn More
Fisherbrand™ 3 Level Flex Rack
Product Code. 17147874 Shop All Fisherbrand Products
Click to view available options
:
Flex Rack
Product Code. 17147874
Only null left
Add to basket
Add to basket
Flexible rack designed for washing in 3 levels.
  • Fitted with a top load indicator that ensures glassware is not damaged when being pushed into the washing chamber
  • Aligned with 8 wheels for smooth, silent, and safe transportation
Specifications
Product Type Flex Rack
For Use With (Application) Washing up to 3 Levels