missing translation for 'onlineSavingsMsg'
Learn More

3-keto-5Beta-Abiraterone, TRC

Product Code. 30393875
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393875 2.5 mg
30403626 25 mg
30408566 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393875 Supplier Toronto Research Chemicals Supplier No. TRCK1709852.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1940175-99-4
Molecular Formula C24H31NO
Molecular Weight (g/mol) 349.51
Synonym (5β)-17-(3-Pyridinyl)androst-16-en-3-one
SMILES C[C@]12CCC(=O)C[C@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC=C4c5cccnc5

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C24H31NO/c1-23-11-9-18(26)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25-15-16)24(21,2)12-10-22(19)23/h3-4,7,13,15,17,19,21-22H,5-6,8-12,14H2,1-2H3/t17-,19+,21+,22+,23+,24-/m1/s1
Chemical Name or Material 3-keto-5Beta-Abiraterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.