CAS RN 1940175-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-keto-5Beta-Abiraterone, TRC
CAS: 1940175-99-4 Molecular Formula: C24H31NO Molecular Weight (g/mol): 349.51 Synonym: (5β)-17-(3-Pyridinyl)androst-16-en-3-one SMILES: C[C@]12CCC(=O)C[C@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC=C4c5cccnc5