missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-L-thyronine, TRC

Product Code. 30010642
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010642 25 mg 25mg
30004106 10 mg 10mg
30009438 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010642 Supplier Toronto Research Chemicals Supplier No. TRCI72150025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10468-90-3
Molecular Formula C15 H14 I N O4
Molecular Weight (g/mol) 399.18
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3-iodophenyl]propanoic acid
SMILES N[C@@H](Cc1ccc(Oc2ccc(O)cc2)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C15H14INO4/c16-12-7-9(8-13(17)15(19)20)1-6-14(12)21-11-4-2-10(18)3-5-11/h1-7,13,18H,8,17H2,(H,19,20)/t13-/m0/s1
Quantity 25 mg
Formula Weight 398.9968
Shipping Condition Room Temperature
Chemical Name or Material 3-Iodo-L-thyronine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.