CAS RN 10468-90-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Iodo-L-thyronine, TRC
CAS: 10468-90-3 Molecular Formula: C15 H14 I N O4 Molecular Weight (g/mol): 399.18 IUPAC Name: (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3-iodophenyl]propanoic acid SMILES: N[C@@H](Cc1ccc(Oc2ccc(O)cc2)c(I)c1)C(=O)O