missing translation for 'onlineSavingsMsg'
Learn More

3-Indolylacetone, TRC

Product Code. 30396490
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396490 250 mg
30393537 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396490 Supplier Toronto Research Chemicals Supplier No. TRCI577350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1201-26-9
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.21
Synonym 1-(1H-Indol-3-yl)-2-propanone, 1-Indol-3-yl-2-propanone, Indol-3-yl-2-propanone, (1H-Indol-3-yl)acetone, 1-(1H-Indol-3-yl)propan-2-one, 3-(2-Oxopropyl)indole, 3-Acetonylindole, 3-Indolylacetone, Indol-3-yl-2-propanone, Indole-3-acetone, NSC 100751
IUPAC Name 1-(1H-indol-3-yl)propan-2-one
SMILES CC(=O)Cc1c[nH]c2ccccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H11NO/c1-8(13)6-9-7-12-11-5-3-2-4-10(9)11/h2-5,7,12H,6H2,1H3
Formula Weight 173.0841
Chemical Name or Material 3-Indolylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.