accessibility menu, dialog, popup

UserName

H3COHN

CAS RN 1201-26-9

IUPAC Name: 1-(1H-indol-3-yl)propan-2-one
Synonyms: 3-indolylacetone
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: LDVHYJKRIKBISQ-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results