missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyflavone, 98+%

Product Code. 11439266
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439266 1 g 1g
11449266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439266 Supplier Thermo Scientific Alfa Aesar Supplier No. A18110.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-85-5 | C15H10O3 | 238.24 g/mol

3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

Solubility
Insoluble in water. Soluble in N,N-DMF and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-85-5
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.24
MDL Number MFCD00006832
InChI Key HVQAJTFOCKOKIN-UHFFFAOYSA-N
Synonym 3-hydroxyflavone, flavonol, 3-hydroxy-2-phenyl-4h-chromen-4-one, flavon-3-ol, 4h-1-benzopyran-4-one, 3-hydroxy-2-phenyl, flavone, 3-hydroxy, 3-hydroxy-2-phenylchromone, unii-ztg9lss5qh, flavone,3-hydroxy, 3-hf
PubChem CID 11349
ChEBI CHEBI:5078
IUPAC Name 3-hydroxy-2-phenylchromen-4-one
SMILES OC1=C(OC2=CC=CC=C2C1=O)C1=CC=CC=C1

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Beilstein 15789
Solubility Information Insoluble in water. Soluble in N,N-DMF and ethanol.
Formula Weight 238.25
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.