missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyagomelatine, TRC

Product Code. 30394001
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394001 5 mg -
30407893 1 mg 1mg
30393314 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394001 Supplier Toronto Research Chemicals Supplier No. TRCH7982805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166526-99-4
Molecular Formula C15H17NO3
Molecular Weight (g/mol) 259.3
Synonym S 21540, N-[2-(3-Hydroxy-7-methoxy-1-naphthalenyl)ethyl]acetamide, Agomelatine 3-Hydroxy Impurity
IUPAC Name N-[2-(3-hydroxy-7-methoxynaphthalen-1-yl)ethyl]acetamide
SMILES OC1=CC(CCNC(C)=O)=C2C(C=CC(OC)=C2)=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H17NO3/c1-10(17)16-6-5-12-8-13(18)7-11-3-4-14(19-2)9-15(11)12/h3-4,7-9,18H,5-6H2,1-2H3,(H,16,17)
Formula Weight 259.1208
Percent Purity >95%
Chemical Name or Material 3-Hydroxyagomelatine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.