CAS RN 166526-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxyagomelatine, TRC
CAS: 166526-99-4 Molecular Formula: C15H17NO3 Molecular Weight (g/mol): 259.3 Synonym: S 21540,N-[2-(3-Hydroxy-7-methoxy-1-naphthalenyl)ethyl]acetamide,Agomelatine 3-Hydroxy Impurity IUPAC Name: N-[2-(3-hydroxy-7-methoxynaphthalen-1-yl)ethyl]acetamide SMILES: OC1=CC(CCNC(C)=O)=C2C(C=CC(OC)=C2)=C1