missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Valproic Acid (Mixture of Diastereomers), TRC

Product Code. 30396046
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396046 5 mg -
30389337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396046 Supplier Toronto Research Chemicals Supplier No. TRCH9930005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58888-84-9
Molecular Formula C8 H16 O3
Molecular Weight (g/mol) 160.21
Synonym 3-Hydroxy Valproic Acid
IUPAC Name 3-hydroxy-2-propylpentanoic acid
SMILES CCCC(C(O)CC)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H16O3/c1-3-5-6(8(10)11)7(9)4-2/h6-7,9H,3-5H2,1-2H3,(H,10,11)
Formula Weight 160.1099
Percent Purity >95%
Chemical Name or Material 3-Hydroxy Valproic Acid (Mixture of Diastereomers)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.