CAS RN 58888-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxy Valproic Acid (Mixture of Diastereomers), TRC
CAS: 58888-84-9 Molecular Formula: C8 H16 O3 Molecular Weight (g/mol): 160.21 Synonym: 3-Hydroxy Valproic Acid IUPAC Name: 3-hydroxy-2-propylpentanoic acid SMILES: CCCC(C(O)CC)C(=O)O