missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-(trifluoromethyl)benzaldehyde (~90%), TRC

Product Code. 30403813
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403813 100 mg 100mg
30398751 50 mg -
30389050 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403813 Supplier Toronto Research Chemicals Supplier No. TRCH954408100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1243282-07-6
Molecular Formula C8H5O2F3
Molecular Weight (g/mol) 190.12
IUPAC Name 3-hydroxy-4-(trifluoromethyl)benzaldehyde
SMILES Oc1cc(C=O)ccc1C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H5F3O2/c9-8(10,11)6-2-1-5(4-12)3-7(6)13/h1-4,13H
Formula Weight 190.0242
Chemical Name or Material 3-Hydroxy-4-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.