CAS RN 1243282-07-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxy-4-(trifluoromethyl)benzaldehyde (~90%), TRC
CAS: 1243282-07-6 Molecular Formula: C8H5O2F3 Molecular Weight (g/mol): 190.12 IUPAC Name: 3-hydroxy-4-(trifluoromethyl)benzaldehyde SMILES: Oc1cc(C=O)ccc1C(F)(F)F