missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%

Product Code. 11448723
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448723 5 g 5g
11438723 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448723 Supplier Thermo Scientific Alfa Aesar Supplier No. A10482.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 537-73-5 | C10H10O4 | 194.186 g/mol

3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-73-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004391
InChI Key QURCVMIEKCOAJU-HWKANZROSA-N
Synonym isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
PubChem CID 736186
ChEBI CHEBI:27794
IUPAC Name (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)O

Specifications

Melting Point ∼231°C (decomposition)
Quantity 5 g
Beilstein 2212760
Solubility Information Soluble in ethanol and methanol.
Formula Weight 194.19
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.