missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic Acid (Isoferulic Acid), TRC

Product Code. 17950325
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950325 500 mg 500mg
17900575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950325 Supplier Toronto Research Chemicals Supplier No. TRCH946180500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-73-5
Molecular Formula C10 H10 O4
Molecular Weight (g/mol) 194.18
Synonym 3-(3-Hydroxy-4-methoxyphenyl)-2-propenoic Acid, Cinnamic acid, 3-hydroxy-4-methoxy- (7CI,8CI), 3-(3-Hydroxy-4-methoxyphenyl)-2-propenoic acid, 3-Hydroxy-4-methoxycinnamic acid, 4-Methoxycaffeic acid, 4-O-Methylcaffeic acid, Hesperetic acid, Isoferulic acid, NSC 51987
IUPAC Name 3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
SMILES COc1ccc(C=CC(=O)O)cc1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H10O4/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)
Formula Weight 194.0579
Chemical Name or Material Isoferulic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.