missing translation for 'onlineSavingsMsg'
Learn More

3-Hexanone, 98%

Product Code. 10791735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791735 250 mL Glass bottle 250mL
10284760 10 mL Glass bottle 10mL
10041600 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10791735 Supplier Thermo Scientific Acros Supplier No. 146902500
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 589-38-8 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-38-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009402
InChI Key PFCHFHIRKBAQGU-UHFFFAOYSA-N
Synonym 3-hexanone, ethyl propyl ketone, 3-oxohexane, aethylpropylketon, ethyl n-propyl ketone, 3-hexanone natural, aethylpropylketon german, fema no. 3290, 3-hexanone, analytical standard, pentanone, methyl
PubChem CID 11509
IUPAC Name hexan-3-one
SMILES CCCC(=O)CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 123°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH2CH2COC2H5
Refractive Index 1.3990 to 1.401
Quantity 250 mL
Beilstein 01, 690
Specific Gravity 0.81
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Hexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.