accessibility menu, dialog, popup

UserName

CAS RN 589-38-8

IUPAC Name: hexan-3-one
Synonyms: 3-hexanone 3-hexanone (natural) aethylpropylketon
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: PFCHFHIRKBAQGU-UHFFFAOYSA-N
SMILES: CCCC(=O)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results