missing translation for 'onlineSavingsMsg'
Learn More

3-Heptyne, 97%

Artikelnummer. 15440398
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15440398 5 g 5g
15450398 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15440398 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H53504.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 2586-89-2 | C7H12 | 96.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2586-89-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00039960
InChI Key KLYHSJRCIZOUHE-UHFFFAOYSA-N
Synonym 3-heptyne, 1-ethyl-2-propylacetylene, acmc-20an2z
PubChem CID 75755
IUPAC Name hept-3-yne
SMILES CCCC#CCC

Tekniske data

Density 0.741
Boiling Point 105°C to 106°C
Flash Point −16°C (−3°F)
Linear Formula CH3(CH2)2C≡CCH2CH3
Refractive Index 1.4225
Quantity 5 g
UN Number UN3295
Beilstein 1731759
Formula Weight 96.17
Percent Purity 97%
Chemical Name or Material 3-Heptyne
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.