accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 2586-89-2

IUPAC Name: hept-3-yne
Synonyms: 3-heptyne
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: KLYHSJRCIZOUHE-UHFFFAOYSA-N
SMILES: CCCC#CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results