missing translation for 'onlineSavingsMsg'
Learn More

3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11452090
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11452090 5 g 5g
11442090 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11452090

Brand: Thermo Scientific Alfa Aesar H28599.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-63-5
Molecular Formula C18H22BNO4
Molecular Weight (g/mol) 327.187
MDL Number MFCD09266173
InChI Key CWNRULQRLNZAGF-UHFFFAOYSA-N
Synonym 3-furfurylaminocarbonyl benzeneboronic acid pinacol ester, n-furan-2-ylmethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, 3-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-yl methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb028, 3-furfurylaminocarbonyl phenylboronic acid, pinacol ester, n-furfuryl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-furan-2-ylmethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
PubChem CID 46739035
IUPAC Name N-(furan-2-ylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)NCC3=CC=CO3

Specifications

Melting Point 122°C to 124°C
Quantity 5 g
Formula Weight 327.18
Percent Purity 97%
Chemical Name or Material 3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.