accessibility menu, dialog, popup

UserName

CAS RN 1073353-63-5

BOOH3CCH3H3CH3COHNO

CAS RN 1073353-63-5

IUPAC Name: N-[(furan-2-yl)methyl]-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-(furan-2-ylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 327.19
Molecular Formula: C18H22BNO4
InChi Key: CWNRULQRLNZAGF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C(=O)NCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results