missing translation for 'onlineSavingsMsg'
Learn More

3-Furaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Product Code. 11368306
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368306 5 g 5g
11358306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368306 Supplier Thermo Scientific Alfa Aesar Supplier No. L15519.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216144-29-5
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD01318151
InChI Key MTZMXGCDOYBCPN-UHFFFAOYSA-N
Synonym 3-diethoxymethyl furan, 3-furaldehyde diethyl acetal, furan,3-diethoxymethyl, diethoxy-3-furylmethane, acmc-1ckie
PubChem CID 7006657
IUPAC Name 3-(diethoxymethyl)furan
SMILES CCOC(OCC)C1=COC=C1

Specifications

Refractive Index 1.444
Quantity 5 g
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 3-Furaldehyde diethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.