CAS RN 216144-29-5
IUPAC Name:
3-(diethoxymethyl)furan
Synonyms:
3-(diethoxymethyl)furan
Molecular Weight (g/mol):
170.21
Molecular Formula:
C9H14O3
InChi Key:
MTZMXGCDOYBCPN-UHFFFAOYSA-N
SMILES:
CCOC(OCC)C1=COC=C1
1
–
1
of
1
results
1
3-Furaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
CAS: 216144-29-5 Molecular Formula: C9H14O3 Molecular Weight (g/mol): 170.21 MDL Number: MFCD01318151 InChI Key: MTZMXGCDOYBCPN-UHFFFAOYSA-N Synonym: 3-diethoxymethyl furan,3-furaldehyde diethyl acetal,furan,3-diethoxymethyl,diethoxy-3-furylmethane,acmc-1ckie PubChem CID: 7006657 IUPAC Name: 3-(diethoxymethyl)furan SMILES: CCOC(OCC)C1=COC=C1