missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)phenol, 97+%, Thermo Scientific™

Product Code. 11450408
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450408 10 g 10g
11440408 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450408 Supplier Thermo Scientific Alfa Aesar Supplier No. B24565.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172333-87-8
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00061288
InChI Key VNTKPYNBATZMSV-UHFFFAOYSA-N
PubChem CID 2737587
IUPAC Name 3-fluoro-5-(trifluoromethyl)phenol
SMILES OC1=CC(F)=CC(=C1)C(F)(F)F

Specifications

Quantity 10 g
UN Number UN3265
Synonym 3-fluoro-5-trifluoromethyl phenol, 5-fluoro-3-trifluoromethyl phenol, phenol, 3-fluoro-5-trifluoromethyl, 3-fluoranyl-5-trifluoromethyl phenol, acmc-1c2u3, 3-fluoro-5-trifluoromethyl-phenol, phenol,3-fluoro-5-trifluoromethyl, 5-fluoro-alpha,alpha,alpha-trifluoro-m-cresol
Formula Weight 180.1
Percent Purity ≥97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.