accessibility menu, dialog, popup

UserName

OHFFFF

CAS RN 172333-87-8

IUPAC Name: 3-fluoro-5-(trifluoromethyl)phenol
Synonyms: 3-fluoro-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: VNTKPYNBATZMSV-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results