missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15467686
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15467686

Brand: Thermo Scientific Alfa Aesar H62652.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 710348-94-0
Molecular Formula C12H15BFNO4
Molecular Weight (g/mol) 267.063
InChI Key CKBJFSWEKLWHTI-UHFFFAOYSA-N
Synonym 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 59770452
IUPAC Name 2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-nitrobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.