accessibility menu, dialog, popup

UserName

CAS RN 710348-94-0

BOOH3CCH3H3CH3CFNOO

CAS RN 710348-94-0

IUPAC Name: 2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 267.06
Molecular Formula: C12H15BFNO4
InChi Key: CKBJFSWEKLWHTI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results