missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-5'-methylacetophenone, 97%, Thermo Scientific™

Product Code. 11481180
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11481180 1 g 1g
11491180 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11481180

Brand: Thermo Scientific Alfa Aesar H26316.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-73-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD04115883
InChI Key KFJMZDKKROJRHE-UHFFFAOYSA-N
Synonym 3'-fluoro-5'-methylacetophenone, 1-3-fluoro-5-methylphenyl ethanone, 1-3-fluoro-5-methylphenyl ethan-1-one, 1-acetyl-3-fluoro-5-methylbenzene
PubChem CID 3803613
IUPAC Name 1-(3-fluoro-5-methylphenyl)ethanone
SMILES CC(=O)C1=CC(C)=CC(F)=C1

Specifications

Refractive Index 1.509
Quantity 1 g
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 3'-Fluoro-5'-methylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.